Malathion (1,2-Bis(ethoxycarbonyl)ethyl-O,O-dimethyldithiophosphat)

Lieferant: LGC Standards PROMOCHEM
Achtung

LGC1205
PROOLGC1205EA 144 EUR
PROOLGC1205
Malathion (1,2-Bis(ethoxycarbonyl)ethyl-O,O-dimethyldithiophosphat)
Malathion (1,2-Bis(ethoxycarbonyl)ethyl-O,O-dimethyldithiophosphat)
Formel: C₁₀H₁₉O₆PS₂
CAS-Nummer: 121-75-5
EINECS: 204-497-7
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR