Aluminium
Lieferant: LGC Standards PROMOCHEM
Synonyme:
Aluminum
LGC2612
PROOLGC2612EA
457
EUR
PROOLGC2612
Aluminium
Aluminium
Formel:
Al Molecular Weight: 26,98 g/mol Siedepunkt: 2327 °C (1013 hPa) Schmelzpunkt: 660 °C Dichte: 2,7 g/cm³ (20 °C) |
MDL:
MFCD00134029 CAS-Nummer: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...