Ketoprofen

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2-(3-Benzoylphenyl)propionic acid, 2-(3-Benzoylphenyl)propanoic acid

MM0001.00
PROOMM0001.00EA 1170 EUR
PROOMM0001.00
Ketoprofen
Ketoprofen
Formel: C₁₆H₁₄O₃
Molecular Weight: 254,29 g/mol
MDL: MFCD00055790
CAS-Nummer: 22071-15-4
EINECS: 244-759-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR