4'-Isobutylacetophenon

Lieferant: LGC Standards PROMOCHEM

Synonyme: 1-(4-Isobutylphenyl)ethanon, 4-Isobutyl-1-acetylbenzol, p-Isobutylacetophenon

MM0002.04
PROOMM0002.04EA 1020 EUR
PROOMM0002.04
4'-Isobutylacetophenon
4'-Isobutylacetophenon
Formel: C₁₂H₁₆O
Molecular Weight: 176.26
MDL: MFCD00027393
CAS-Nummer: 38861-78-8
EINECS: 254-159-8
UN: 1224
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR