4'-Isopropylacetophenon
Lieferant: LGC Standards PROMOCHEM
Synonyme:
1-(4-Isopropylphenyl)ethanon, 4-Isopropyl-1-acetylbenzol, p-Isopropylacetophenon
MM0002.57
PROOMM0002.57EA
1830
EUR
PROOMM0002.57
4'-Isopropylacetophenon
4'-Isopropylacetophenon
Formel:
C₁₁H₁₄O Molecular Weight: 162,23 g/mol Dichte: 0,97 g/cm³ (20 °C) |
MDL:
MFCD00048297 CAS-Nummer: 645-13-6 EINECS: 211-433-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...