trans-4-Aminocyclohexanol Hydrochlorid

Lieferant: LGC Standards PROMOCHEM

MM0005.06
PROOMM0005.06EA 301 EUR
PROOMM0005.06
trans-4-Aminocyclohexanol Hydrochlorid
trans-4-Aminocyclohexanol Hydrochlorid
Formel: C₆H₁₄ClNO
Molecular Weight: 151,64 g/mol
MDL: MFCD00012566
CAS-Nummer: 50910-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR