Diclofenac-Natrium

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt, GP-45840, [o-(2,6-dichloroanilino) phenyl]acetic acid sodium salt, 2-[(2,6-Dichlorophenyl)amino]-benzeneacetic acid monosodium salt, Sodium [o-[(2,6-dichlorophenyl) amino] phenyl] acetate

MM0006.00
PROOMM0006.00EA 1160 EUR
PROOMM0006.00
Diclofenac-Natrium
Diclofenac-Natrium
Formel: C₁₄H₁₀Cl₂NNaO₂
Molecular Weight: 318,1 g/mol
MDL: MFCD00082251
CAS-Nummer: 15307-79-6
EINECS: 239-346-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR