(4-Hydroxyphenyl)essigsäure
Lieferant: LGC Standards PROMOCHEM
|
Achtung
|
Synonyme:
4-Hydroxyphenylacetic Acid
MM0013.13
PROOMM0013.13EA
598
EUR
PROOMM0013.13
(4-Hydroxyphenyl)essigsäure
(4-Hydroxyphenyl)essigsäure
Formel:
(C₆H₄(OH))CH₂COOH Molecular Weight: 152,15 g/mol Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: 147…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004347 CAS-Nummer: 156-38-7 EINECS: 205-851-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...