Pyridoxal Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
Synonyme:
3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl
MM0021.05
PROOMM0021.05EA
1330
EUR
PROOMM0021.05
Pyridoxal Hydrochlorid
Pyridoxal Hydrochlorid
Formel:
C₈H₁₀ClNO₃ Molecular Weight: 203,62 g/mol |
MDL:
MFCD00012809 CAS-Nummer: 65-22-5 EINECS: 200-602-5 |