(3,4-Dimethoxyphenyl)acetonitril

Lieferant: LGC Standards PROMOCHEM

Synonyme: Homoveratronitril, 3,4-Dimethoxybenzylcyanid

MM0022.18
PROOMM0022.18EA 935 EUR
PROOMM0022.18
(3,4-Dimethoxyphenyl)acetonitril
(3,4-Dimethoxyphenyl)acetonitril
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Siedepunkt: 320 °C (1013 hPa)
Schmelzpunkt: 60…62 °C
MDL: MFCD00001911
CAS-Nummer: 93-17-4
EINECS: 202-225-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR