2'-Benzoyl-2,4'-dichloracetanilid

Lieferant: LGC Standards PROMOCHEM

Synonyme: N1-(2-Benzoyl-4-chlorophenyl)-2-chloroacetamide, N-(2-Benzoyl-4-chlorophenyl)-2-chloroacetamide

MM0023.05
PROOMM0023.05EA 598 EUR
PROOMM0023.05
2'-Benzoyl-2,4'-dichloracetanilid
2'-Benzoyl-2,4'-dichloracetanilid
Formel: C₁₅H₁₁Cl₂NO₂
Molecular Weight: 308,159 g/mol
MDL: MFCD00018905
CAS-Nummer: 4016-85-7
EINECS: 223-678-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR