4'-Hydroxyacetophenonoxim

Lieferant: LGC Standards PROMOCHEM

MM0042.12
PROOMM0042.12EA 1220 EUR
PROOMM0042.12
4'-Hydroxyacetophenonoxim
4'-Hydroxyacetophenonoxim
Formel: C₈H₉NO₂
MDL: MFCD00481697
CAS-Nummer: 34523-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR