Thiamin Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
Synonyme:
Thiaminiumchlorid, Antiberiberifaktor Hydrochlorid, antineuritisches Vitamin Hydrochlorid, Aneurin Hydrochlorid, Thiaminchlorid Hydrochlorid, Vitamin B1 Hydrochlorid
MM0049.00
PROOMM0049.00EA
1450
EUR
PROOMM0049.00
Thiamin Hydrochlorid
Thiamin Hydrochlorid
Formel:
C₁₂H₁₈Cl₂N₄OS Molecular Weight: 337.27 g/mol Schmelzpunkt: 252 Dichte: 1.401 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012780 CAS-Nummer: 67-03-8 EINECS: 200-641-8 Merck Index: 13,09366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...