2-(4-Methyl-1,3-thiazol-5-yl)ethanol
Lieferant: LGC Standards PROMOCHEM
Synonyme:
5-(2-Hydroxyethyl)-4-methylthiazol, 4-Methyl-5-(2-hydroxyethyl)thiazol, 4-Methyl-5-thiazoleethanol
MM0049.02
PROOMM0049.02EA
1090
EUR
PROOMM0049.02
2-(4-Methyl-1,3-thiazol-5-yl)ethanol
2-(4-Methylthiazol-5-yl)ethanol
Formel:
C₆H₉NOS Molecular Weight: 143,21 g/mol Siedepunkt: 135 °C (1013 hPa) Dichte: 1,199 g/cm³ (20 °C) |
MDL:
MFCD00005339 CAS-Nummer: 137-00-8 EINECS: 205-272-6 Merck Index: 12,06208 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...