1,1-Dimethylbiguanid Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
Synonyme:
AMPK Signaling Activator VII, Metformin, Metformin hydrochloride, 1,1-Dimethylbiguanide Hydrochloride
MM0056.00
PROOMM0056.00EA
1170
EUR
PROOMM0056.00
1,1-Dimethylbiguanid Hydrochlorid
1,1-Dimethylbiguanid Hydrochlorid
Formel:
C₄H₁₂ClN₅ Molecular Weight: 165.625 g/mol |
MDL:
MFCD00012582 CAS-Nummer: 1115-70-4 EINECS: 214-230-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...