Melamin

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2,4,6-Triamino-1,3,5-triazin, 2,4,6-Triamino-s-triazin

MM0056.04
PROOMM0056.04EA 418 EUR
PROOMM0056.04
Melamin
Melamin
Formel: C₃H₆N₆
Molecular Weight: 126,12 g/mol
Dichte: 1,57 g/cm³ (20 °C)
Flammpunkt: >280 °C
MDL: MFCD00006055
CAS-Nummer: 108-78-1
EINECS: 203-615-4
Merck Index: 13,05834

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR