Dibenzo[a,d]cycloheptan
Lieferant: LGC Standards PROMOCHEM
Synonyme:
10,11-Dihydro-5H-dibenzo[a,d]cyclohepten
MM0062.13
PROOMM0062.13EA
425
EUR
PROOMM0062.13
Dibenzo[a,d]cycloheptan
Dibenzo[a,d]cycloheptan
Formel:
C₁₅H₁₄ Molecular Weight: 194,28 g/mol Siedepunkt: 357 °C (1013 hPa) Schmelzpunkt: 78 °C Dichte: 1,19 g/cm³ (20 °C) |
CAS-Nummer:
833-48-7 EINECS: 212-630-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...