Tyramin

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2-(4-Hydroxyphenyl)ethylamin, 4-Hydroxyphenethylamim, 4-(2-Aminoethyl)phenol

MM0063.06
PROOMM0063.06EA 685 EUR
PROOMM0063.06
Tyramin
Tyramin
Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Siedepunkt: 325 °C (1013 hPa)
Schmelzpunkt: 164 °C
MDL: MFCD00008193
CAS-Nummer: 51-67-2
EINECS: 200-115-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR