2-Amino-5-brombenzoylpyridin
Lieferant: LGC Standards PROMOCHEM
Synonyme:
(2-Amino-5-bromophenyl)(pyridin-2-yl)methanone, 2-(2-Amino-5-bromobenzoyl)pyridine
MM0064.01
PROOMM0064.01EA
1320
EUR
PROOMM0064.01
2-Amino-5-brombenzoylpyridin
2-Amino-5-brombenzoylpyridin
Formel:
C₁₂H₉BrN₂O Molecular Weight: 277,1167 g/mol |
MDL:
MFCD07780283 CAS-Nummer: 1563-56-0 EINECS: 216-352-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...