Phloroglucintrimethylether

Lieferant: LGC Standards PROMOCHEM

Synonyme: O,O,O-1,3,5-Trimethylresorcin, 1,3,5-Trimethoxybenzol

MM0066.05
PROOMM0066.05EA 664 EUR
PROOMM0066.05
Phloroglucintrimethylether
Phloroglucintrimethylether
Formel: C₉H₁₂O₃
Molecular Weight: 168,19 g/mol
Siedepunkt: 255 °C (1013 hPa)
Schmelzpunkt: 52…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008385
CAS-Nummer: 621-23-8
EINECS: 210-673-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR