(S)-(-)-3-Acetylthio-2-methylpropionsäure

Lieferant: LGC Standards PROMOCHEM

MM0067.04
PROOMM0067.04EA 674 EUR
PROOMM0067.04
(S)-(-)-3-Acetylthio-2-methylpropionsäure
(S)-(-)-3-Acetylthio-2-methylpropionsäure
Formel: C₆H₁₀O₃S
Molecular Weight: 162,21 g/mol
CAS-Nummer: 76497-39-7
EINECS: 278-480-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR