1-Vinyl-2-pyrrolidon

Lieferant: LGC Standards PROMOCHEM

Synonyme: N-Vinyl-2-pyrrolidone, 1-Vinyl-2-pyrrolidinone, N-Vinyl-2-pyrrolidinone

MM0083.03
PROOMM0083.03EA 1200 EUR
PROOMM0083.03
1-Vinyl-2-pyrrolidon
1-Vinyl-2-pyrrolidon
Formel: C₆H₉NO
Molecular Weight: 111,1418 g/mol
MDL: MFCD00003197
CAS-Nummer: 88-12-0
EINECS: 201-800-4
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR