Ethyl-4-nitrobenzoat

Lieferant: LGC Standards PROMOCHEM

Synonyme: 4-Nitrobenzoesäureethylester

MM0094.04
PROOMM0094.04EA 493 EUR
PROOMM0094.04
Ethyl-4-nitrobenzoat
Ethyl-4-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007351
CAS-Nummer: 99-77-4
EINECS: 202-786-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR