2-Chlor-N-(2,6-dimethylphenyl)acetamid
Lieferant: LGC Standards PROMOCHEM
Synonyme:
N-(2,6-Dimethylphenyl)chloroacetamide, 2-Chloro-2',6'-dimethylacetanilide, N-(2,6-Dichlorophenyl)-2-chloroacetamide
MM0102.03
PROOMM0102.03EA
677
EUR
PROOMM0102.03
2-Chlor-N-(2,6-dimethylphenyl)acetamid
2-Chlor-N-(2,6-dimethylphenyl)acetamid
Formel:
C₁₀H₁₂ClNO Molecular Weight: 197,66 g/mol |
MDL:
MFCD00000926 CAS-Nummer: 1131-01-7 EINECS: 214-460-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...