4-Chlor-4'-fluorbutyrophenon
Lieferant: LGC Standards PROMOCHEM
Synonyme:
γ-Chlor-4-fluorbutyrophenon, ω-Chlor-4-fluorbutyrophenon
MM0113.07
PROOMM0113.07EA
531
EUR
PROOMM0113.07
4-Chlor-4'-fluorbutyrophenon
4-Chlor-4'-fluorbutyrophenon
Formel:
C₁₀H₁₀ClFO Molecular Weight: 200,64 g/mol Siedepunkt: 300 °C (1013 hPa) Dichte: 1,2267 g/cm³ (20 °C) |
MDL:
MFCD00001007 CAS-Nummer: 3874-54-2 EINECS: 223-395-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...