Isosorbiddimethylether

Lieferant: LGC Standards PROMOCHEM

Synonyme: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan

MM0126.02
PROOMM0126.02EA 809 EUR
PROOMM0126.02
Isosorbiddimethylether
Isosorbiddimethylether
Formel: C₈H₁₄O₄
Molecular Weight: 174,20 g/mol
MDL: MFCD00064828
CAS-Nummer: 5306-85-4
EINECS: 226-159-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR