Isosorbid

Lieferant: LGC Standards PROMOCHEM

Synonyme: D-Isosorbide, (3R,3aR,6S,6aR)-Hexahydrofuro[3,2-b]furan-3,6-diol

MM0126.03
PROOMM0126.03EA 1050 EUR
PROOMM0126.03
Isosorbid
Isosorbid
Formel: C₆H₁₀O₄
Molecular Weight: 146,14 g/mol
MDL: MFCD00064827
CAS-Nummer: 652-67-5
EINECS: 211-492-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR