Estriol
Lieferant: LGC Standards PROMOCHEM
MM0130.00
PROOMM0130.00EA
1230
EUR
PROOMM0130.00
Estriol
Estriol
Formel:
C₁₈H₂₄O₃ |
MDL:
MFCD00003691 CAS-Nummer: 50-27-1 EINECS: 200-022-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...