4-Chlorphenylharnstoff
Lieferant: LGC Standards PROMOCHEM
Synonyme:
1-(4-Chlorophenyl)urea
MM0162.07
PROOMM0162.07EA
1320
EUR
PROOMM0162.07
4-Chlorphenylharnstoff
4-Chlorphenylharnstoff
Formel:
C₇H₇ClN₂O Molecular Weight: 170,6 g/mol |
MDL:
MFCD00014788 CAS-Nummer: 140-38-5 EINECS: 205-412-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...