cis-Anethol

Lieferant: LGC Standards PROMOCHEM

MM0164.06
PROOMM0164.06EA 3590 EUR
PROOMM0164.06
cis-Anethol
cis-Anethol
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
CAS-Nummer: 25679-28-1
EINECS: 247-181-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR