2-Butyl-5-chlorimidazol-4-carbaldehyd

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2-Butyl-4-chloro-5-formylimidazole, 2-(But-1-yl)-5-chloro-1H-imidazole-4-carboxaldehyde, 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde

MM0168.08
PROOMM0168.08EA 1020 EUR
PROOMM0168.08
2-Butyl-5-chlorimidazol-4-carbaldehyd
2-Butyl-5-chlorimidazol-4-carbaldehyd
Formel: C₈H₁₁ClN₂O
Molecular Weight: 186,6387 g/mol
MDL: MFCD01934396
CAS-Nummer: 83857-96-9
EINECS: 410-260-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR