N-Acetyl-DL-Tryptophan

Lieferant: LGC Standards PROMOCHEM

MM0172.13
PROOMM0172.13EA 838 EUR
PROOMM0172.13
N-Acetyl-DL-Tryptophan
N-Acetyl-DL-Tryptophan
Formel: C₁₃H₁₄N₂O₃
Schmelzpunkt: 205…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005644
CAS-Nummer: 87-32-1
EINECS: 201-739-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR