3-Methylxanthin
Lieferant: LGC Standards PROMOCHEM
|
Achtung
|
Synonyme:
3-Methyl-1H,3H-purin-2,6-dion, 3-Methyl-2,6(1H,3H)-purindion, 3-Methyl-3,7-dihydro-1H-purin-2,6-dion, 3,7-Dihydro-3-methyl-1H-purin-2,6-dion, 2,6-Dihydroxy-3-methylpurin, 3-Methyl-3,7-dihydro-2H-purin-2,6-dion
MM0176.03
PROOMM0176.03EA
1020
EUR
PROOMM0176.03
3-Methylxanthin
3-Methylxanthin
Formel:
C₆H₆N₄O₂ Molecular Weight: 166,14 g/mol |
MDL:
MFCD00005580 CAS-Nummer: 1076-22-8 EINECS: 214-058-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...