Benzyldimethyldodecylammoniumchlorid

Lieferant: LGC Standards PROMOCHEM

Synonyme: Benzyldimethyldodecylammonium chloride, N-Benzyl-N,N-dimethyldodecan-1-aminium chloride, Dodecyldimethylbenzylammonium chloride, Benzyldodecyldimethylammonium chloride dihydrate

MM0224.01
PROOMM0224.01EA 1390 EUR
PROOMM0224.01
Benzyldimethyldodecylammoniumchlorid
Benzyldimethyldodecylammoniumchlorid
Formel: C₂₁H₃₈ClN
Molecular Weight: 339,99 g/mol
MDL: MFCD00137276
CAS-Nummer: 139-07-1
EINECS: 205-351-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR