Benzyldimethylhexadecylammoniumchlorid

Lieferant: LGC Standards PROMOCHEM

Synonyme: Cetalkonium chloride, N-Benzyl-N,N-dimethylhexadecan-1-aminium chloride

MM0224.03
PROOMM0224.03EA 1830 EUR
PROOMM0224.03
Benzyldimethylhexadecylammoniumchlorid
Benzyldimethylhexadecylammoniumchlorid
Formel: C₂₅H₄₆ClN
Molecular Weight: 396,10 g/mol
MDL: MFCD00011625
CAS-Nummer: 122-18-9
EINECS: 204-526-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR