3-Amino-5-methylisoxazol

Lieferant: LGC Standards PROMOCHEM

Synonyme: 5-Methylisoxazol-3-ylamin, 5-Methylisoxazol-3-amin

MM0227.02
PROOMM0227.02EA 422 EUR
PROOMM0227.02
3-Amino-5-methylisoxazol
3-Amino-5-methylisoxazole
Formel: C₄H₆N₂O
Molecular Weight: 98,1 g/mol
Siedepunkt: 194 °C (1013 hPa)
Schmelzpunkt: 56…60 °C
MDL: MFCD00003155
CAS-Nummer: 1072-67-9
EINECS: 214-013-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR