1,2,4-Triazol
Lieferant: LGC Standards PROMOCHEM
|
Achtung
|
Synonyme:
1,2,4-1H-Triazol
MM0235.03
PROOMM0235.03EA
563
EUR
PROOMM0235.03
1,2,4-Triazol
1,2,4-Triazole
Formel:
C₂H₃N₃ Molecular Weight: 69.07 g/mol Siedepunkt: 260 Schmelzpunkt: 116…119 °C Dichte: 1.394 g/cm³ (25 °C) |
MDL:
MFCD00005228 CAS-Nummer: 288-88-0 EINECS: 206-022-9 Merck Index: 12,09735 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...