ω-Chlor-2,4-difluoracetophenon

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2-Chlor-2',4'-difluoracetophenon, α-Chlor-2,4-difluoracetophenon

MM0235.04
PROOMM0235.04EA 679 EUR
PROOMM0235.04
ω-Chlor-2,4-difluoracetophenon
ω-Chlor-2,4-difluoracetophenon
Formel: C₈H₅ClF₂O
Molecular Weight: 190,58 g/mol
Siedepunkt: 233 °C (1013 hPa)
Schmelzpunkt: 47…48 °C
MDL: MFCD00013252
CAS-Nummer: 51336-94-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR