4-Amino-4H-1,2,4-triazol

Lieferant: LGC Standards PROMOCHEM

Synonyme: 4-Amino-1,2,4-triazol, 1,2,4-Triazol-4-ylamin

MM0235.09
PROOMM0235.09EA 686 EUR
PROOMM0235.09
4-Amino-4H-1,2,4-triazol
4-Amino-4H-1,2,4-triazol
Formel: C₂H₄N₄
Molecular Weight: 84,08 g/mol
MDL: MFCD00003099
CAS-Nummer: 584-13-4
EINECS: 209-533-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR