Ampicillin Trihydrat

Lieferant: LGC Standards PROMOCHEM
Achtung

MM0253.15
PROOMM0253.15EA 264 EUR
PROOMM0253.15
Ampicillin Trihydrat
Ampicillin Trihydrat
Formel: C₁₆H₁₉N₃O₄S·3H₂O
Lagertemperatur: Kühlschrank
MDL: MFCD00072036
CAS-Nummer: 7177-48-2
EINECS: 200-709-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR