4-Methylumbelliferon
Lieferant: LGC Standards PROMOCHEM
Synonyme:
7-Hydroxy-4-methylcumarin, Hymecromon
MM0294.00
PROOMM0294.00EA
1360
EUR
PROOMM0294.00
4-Methylumbelliferon
4-Methylumbelliferon
Formel:
C₁₀H₈O₃ Molecular Weight: 176,17 g/mol |
MDL:
MFCD00006866 CAS-Nummer: 90-33-5 EINECS: 201-986-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...