7-Hydroxy-2-methylchromon

Lieferant: LGC Standards PROMOCHEM

Synonyme: 7-Hydroxy-2-methyl-4H-chromen-4-one

MM0294.02
PROOMM0294.02EA 1630 EUR
PROOMM0294.02
7-Hydroxy-2-methylchromon
7-Hydroxy-2-methylchromon
Formel: C₁₀H₈O₃
Molecular Weight: 176,1700 g/mol
MDL: MFCD00208225
CAS-Nummer: 6320-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR