trans-4-(Aminomethyl)cyclohexancarbonsäure

Lieferant: LGC Standards PROMOCHEM

Synonyme: HAKU, AMCA, TAMCHA, trans-4-(aminomethyl)cyclohexane-1-carboxylic acid, Tranexamic acid, AMCHA

MM0330.00
PROOMM0330.00EA 1180 EUR
PROOMM0330.00
trans-4-(Aminomethyl)cyclohexancarbonsäure
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
MDL: MFCD00001466
CAS-Nummer: 1197-18-8
EINECS: 214-818-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR