Guaifenesin
Lieferant: LGC Standards PROMOCHEM
MM0354.00
PROOMM0354.00EA
1230
EUR
PROOMM0354.00
Guaifenesin
Guaifenesin
Formel:
C₁₀H₁₄O₄ |
MDL:
MFCD00016873 CAS-Nummer: 93-14-1 EINECS: 202-222-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...