N-tert-Butylbenzylamin
Lieferant: LGC Standards PROMOCHEM
Synonyme:
N-Benzyl-tert-butylamine, N-Benzyl-2-methylpropan-2-amine
MM0440.03
PROOMM0440.03EA
321
EUR
PROOMM0440.03
N-tert-Butylbenzylamin
N-tert-Butylbenzylamin
Formel:
C₁₁H₁₇N Molecular Weight: 163,26 g/mol |
MDL:
MFCD00008798 CAS-Nummer: 3378-72-1 EINECS: 222-179-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...