Dexamethason-21-acetat

Lieferant: LGC Standards PROMOCHEM

Synonyme: 11β,17α-Dihydroxy-21-(ethanoyloxy)-9-fluor-16α-methyl-1,4-pregnadien-3,20-dion, Dexamethasone acetate

MM0445.00
PROOMM0445.00EA 254 EUR
PROOMM0445.00
Dexamethason-21-acetat
Dexamethason-21-acetat
Formel: C₂₄H₃₁FO₆
Molecular Weight: 434,5 g/mol
MDL: MFCD00027407
CAS-Nummer: 1177-87-3
EINECS: 214-646-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR