2-Hydroxybenzimidazol
Lieferant: LGC Standards PROMOCHEM
Synonyme:
2-Benzimidazolinon, 2(1H)-Benzimidazolon, 2-Hydroxy-1H-benzimidazol
MM0474.03
PROOMM0474.03EA
791
EUR
PROOMM0474.03
2-Hydroxybenzimidazol
2-Hydroxybenzimidazole
Formel:
C₇H₆N₂O Molecular Weight: 134,14 g/mol Schmelzpunkt: 320…322 °C Dichte: 1,42 g/cm³ (20 °C) |
MDL:
MFCD00127894 CAS-Nummer: 615-16-7 EINECS: 210-412-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...