2-Methylindolin

Lieferant: LGC Standards PROMOCHEM
Achtung

MM0477.08
PROOMM0477.08EA 1060 EUR
PROOMM0477.08
2-Methylindolin
2-Methylindolin
Formel: C₉H₁₁N
Molecular Weight: 133,19 g/mol
Siedepunkt: 228 °C (1013 hPa)
Dichte: 1,0236 g/cm³ (15 °C)
MDL: MFCD00005706
CAS-Nummer: 6872-06-6
EINECS: 229-971-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR