Isobutyl-4-hydroxybenzoat

Lieferant: LGC Standards PROMOCHEM

Synonyme: Isobutyl 4-hydroxybenzoate, Isobutylparaben

MM0496.05
PROOMM0496.05EA 1620 EUR
PROOMM0496.05
Isobutyl-4-hydroxybenzoat
Isobutyl-4-hydroxybenzoat
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
MDL: MFCD00020167
CAS-Nummer: 4247-02-3
EINECS: 224-208-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR