Amantadin Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
Synonyme:
1-Adamantylamin Hydrochlorid, 1-Aminotricyclo[3.3.1.1(3,7)]decan Hydrochlorid, Tricyclo[3.3.1.1(3,7)]dec-1-ylaminhydrochlorid, 1-Adamantanammoniumchlorid, 1-Aminoadamantan Hydrochlorid
MM0564.00
PROOMM0564.00EA
1190
EUR
PROOMM0564.00
Amantadin Hydrochlorid
Amantadin Hydrochlorid
Formel:
C₁₀H₁₈ClN Molecular Weight: 187,71 g/mol Schmelzpunkt: 360 °C |
MDL:
MFCD00074723 CAS-Nummer: 665-66-7 EINECS: 211-560-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...